Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sra-33
Entrez Gene ID
181920
Gene Name
Serpentine receptor class alpha-33
Gene Aliases
CELE_B0304.6
Chromosome Location
Chr.II: 4534736 - 4537241 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062401.7 NP_494802.3 1 - 2 117 122

View Full Product Details