Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fut-6
Entrez Gene ID
188084
Gene Name
Alpha-(1,3)-fucosyltransferase fut-6
Gene Aliases
CELE_T05A7.5
Chromosome Location
Chr.II: 4653175 - 4655934 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062422.8 NP_494823.2 4 - 5 463 76
GenBank mRNA AJ745074.1 - 4 - 5 461 76

View Full Product Details