Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-20
Entrez Gene ID
173840
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F11G11.11
Chromosome Location
Chr.II: 4868807 - 4870791 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062478.9 NP_494879.2 1 - 2 87 65

View Full Product Details