Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zyg-12
Entrez Gene ID
24105291
Gene Name
protein-coding
Gene Aliases
CELE_ZK546.1
Chromosome Location
Chr.II: 4949900 - 4953492 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306590.3 NP_001293519.1 1 - 2 168 65
NM_001306591.3 NP_001293520.1 1 - 2 168 65
NM_001306592.3 NP_001293521.1 1 - 2 168 65
NM_001373710.2 NP_001360114.1 1 - 2 166 65
GenBank mRNA AY487140.1 - 1 - 2 164 65

Gene Symbol
ndx-3
Entrez Gene ID
173857
Gene Name
Nudix hydrolase 3
Gene Aliases
CELE_Y38A8.1
Chromosome Location
Chr.II: 4949900 - 4954505 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383822.1 1 - 2 168 65

View Full Product Details