Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- zyg-12
- Entrez Gene ID
-
24105291
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_ZK546.1
- Chromosome Location
-
Chr.II: 4949900 - 4953492 on Build WBcel235
- Gene Symbol
- ndx-3
- Entrez Gene ID
-
173857
- Gene Name
- Nudix hydrolase 3
- Gene Aliases
-
CELE_Y38A8.1
- Chromosome Location
-
Chr.II: 4949900 - 4954505 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383822.1 |
|
4 - 5 |
1500 |
75 |
View Full Product Details