Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsy-1
Entrez Gene ID
24104671
Gene Name
protein-coding
Gene Aliases
CELE_F59A6.1
Chromosome Location
Chr.II: 5022757 - 5029999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313625.4 NP_001300554.1 4 - 5 1054 77
NM_001383826.1 5 - 6 1253 77

View Full Product Details