Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srb-5
Entrez Gene ID
191787
Gene Name
Serpentine receptor class beta-5
Gene Aliases
CELE_C27D6.6
Chromosome Location
Chr.II: 5161995 - 5163383 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062560.1 NP_494961.1 3 - 4 657 153

View Full Product Details