Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pkc-3
Entrez Gene ID
173914
Gene Name
Protein kinase C-like 3
Gene Aliases
CELE_F09E5.1
Chromosome Location
Chr.II: 5380027 - 5384169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062610.9 NP_495011.1 7 - 8 1322 125
GenBank mRNA AB007320.1 - 7 - 8 1341 125
AF025666.1 - 7 - 8 1324 125

View Full Product Details