Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmr-1
Entrez Gene ID
173929
Gene Name
LITAF domain-containing protein
Gene Aliases
CELE_F07F6.6
Chromosome Location
Chr.II: 5440238 - 5446606 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062632.7 NP_495033.2 14 - 15 2213 116
GenBank mRNA AF318613.2 - 14 - 15 2211 116

View Full Product Details