Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cex-1
Entrez Gene ID
186376
Gene Name
Calexcitin-1
Gene Aliases
CELE_F56D1.6
Chromosome Location
Chr.II: 5447681 - 5448634 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062633.2 NP_495034.1 3 - 4 486 69

View Full Product Details