Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifb-2
Entrez Gene ID
173973
Gene Name
protein-coding
Gene Aliases
CELE_F10C1.7
Chromosome Location
Chr.II: 5746731 - 5754143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062732.6 NP_495133.1 6 - 7 1227 59

View Full Product Details