Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptp-2
Entrez Gene ID
24104681
Gene Name
Tyrosine-protein phosphatase non-receptor type ptp-2
Gene Aliases
CELE_F59G1.5
Chromosome Location
Chr.II: 5918647 - 5921617 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306583.2 NP_001293512.1 3 - 4 748 121
GenBank mRNA AF015882.1 - 3 - 4 749 121

View Full Product Details