Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exp-1
Entrez Gene ID
186801
Gene Name
Gamma-aminobutyric acid receptor exp-1
Gene Aliases
CELE_H35N03.1
Chromosome Location
Chr.II: 6157144 - 6161094 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062828.6 NP_495229.3 10 - 11 961 113

View Full Product Details