Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkh-8
Entrez Gene ID
174026
Gene Name
Fork-head domain-containing protein
Gene Aliases
CELE_F40H3.4
Chromosome Location
Chr.II: 6186719 - 6191402 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267178.3 NP_001254107.1 7 - 8 857 120

View Full Product Details