Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ani-2
Entrez Gene ID
174055
Gene Name
Anillin-like protein 2
Gene Aliases
CELE_K10B2.5
Chromosome Location
Chr.II: 6361159 - 6365367 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062881.4 NP_495282.1 1 - 2 176 90

View Full Product Details