Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-23
Entrez Gene ID
174058
Gene Name
protein-coding
Gene Aliases
CELE_K10B2.1
Chromosome Location
Chr.II: 6371303 - 6374776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313634.3 NP_001300563.1 6 - 7 1728 69
NM_062884.7 NP_495285.1 7 - 8 1901 69
GenBank mRNA AF275253.1 - 7 - 8 1908 69

View Full Product Details