Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abch-1
Entrez Gene ID
174078
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_C56E6.5
Chromosome Location
Chr.II: 6528929 - 6531627 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062919.5 NP_495320.1 5 - 6 850 122

View Full Product Details