Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- icd-1
- Entrez Gene ID
-
174090
- Gene Name
- Transcription factor BTF3 homolog
- Gene Aliases
-
CELE_C56C10.8
- Chromosome Location
-
Chr.II: 6584448 - 6588655 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062935.9 |
NP_495336.1 |
1 - 2 |
85 |
106 |
- Gene Symbol
- C56C10.7
- Entrez Gene ID
-
174089
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C56C10.7
- Chromosome Location
-
Chr.II: 6584409 - 6588655 on Build WBcel235
View Full Product Details