Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-8
Entrez Gene ID
174093
Gene Name
protein-coding
Gene Aliases
CELE_C56C10.13
Chromosome Location
Chr.II: 6594370 - 6601074 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047288.4 NP_001040753.1 7 - 8 1389 167
NM_001047289.6 NP_001040754.1 3 - 4 777 167

View Full Product Details