Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-10
Entrez Gene ID
192063
Gene Name
protein-coding
Gene Aliases
CELE_T14B4.4
Chromosome Location
Chr.II: 6718586 - 6720269 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047340.5 NP_001040805.1 5 - 6 482 77
NM_001047341.4 NP_001040806.1 3 - 4 308 77

View Full Product Details