Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-5
Entrez Gene ID
174122
Gene Name
protein-coding
Gene Aliases
CELE_C04A2.7
Chromosome Location
Chr.II: 6853458 - 6858301 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062999.9 NP_495400.2 13 - 14 2550 110

View Full Product Details