Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmp-1
Entrez Gene ID
174126
Gene Name
Peroxisomal Membrane Protein related
Gene Aliases
CELE_C44B7.8
Chromosome Location
Chr.II: 6880699 - 6883754 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063006.6 NP_495407.1 4 - 5 542 150

View Full Product Details