Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-14
Entrez Gene ID
174157
Gene Name
MutS protein homolog him-14
Gene Aliases
CELE_ZK1127.11
Chromosome Location
Chr.II: 7065020 - 7069966 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063050.6 NP_495451.1 12 - 13 1868 77
GenBank mRNA AF178755.1 - 12 - 13 1868 77

View Full Product Details