Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kat-1
Entrez Gene ID
174161
Gene Name
Acetyl-CoA acetyltransferase homolog, mitochondrial
Gene Aliases
CELE_T02G5.8
Chromosome Location
Chr.II: 7073814 - 7075551 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063054.7 NP_495455.2 4 - 5 763 77

View Full Product Details