Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
farl-11
Entrez Gene ID
174165
Gene Name
protein-coding
Gene Aliases
CELE_F10E7.8
Chromosome Location
Chr.II: 7099594 - 7105035 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001346630.2 NP_001333549.1 4 - 5 539 157
NM_063066.8 NP_495467.2 4 - 5 532 157

View Full Product Details