Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spon-1
Entrez Gene ID
174169
Gene Name
SPONdin (extracellular matrix glycoprotein) family
Gene Aliases
CELE_F10E7.4
Chromosome Location
Chr.II: 7116557 - 7120665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063072.8 NP_495473.2 4 - 5 505 64

View Full Product Details