Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-59
Entrez Gene ID
191820
Gene Name
Serpentine receptor class delta-59
Gene Aliases
CELE_E04F6.1
Chromosome Location
Chr.II: 7205688 - 7207038 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063098.2 NP_495499.1 1 - 2 306 87

View Full Product Details