Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul-4
Entrez Gene ID
174198
Gene Name
Cullin-4
Gene Aliases
CELE_F45E12.3
Chromosome Location
Chr.II: 7302101 - 7305765 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063124.7 NP_495525.2 2 - 3 220 73

View Full Product Details