Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-21
Entrez Gene ID
174202
Gene Name
protein-coding
Gene Aliases
CELE_F21D12.1
Chromosome Location
Chr.II: 7339227 - 7349133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026929.4 NP_001022100.1 6 - 7 847 153
NM_001026930.4 NP_001022101.1 5 - 6 1374 153
NM_001026931.6 NP_001022102.1 6 - 7 862 153
NM_001026932.5 NP_001022103.1 6 - 7 820 153
NM_001047301.4 NP_001040766.1 6 - 7 860 153
GenBank mRNA AF083234.1 - 4 - 5 751 153

View Full Product Details