The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001381495.1 | NP_001368642.1 | 9 - 10 | 1037 | 71 |
NM_001381496.2 | NP_001367460.1 | 9 - 10 | 1061 | 71 |