Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-4
Entrez Gene ID
174208
Gene Name
RPOL4c domain-containing protein
Gene Aliases
CELE_F43E2.2
Chromosome Location
Chr.II: 7371771 - 7375187 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383887.1 5 - 6 1785 58

View Full Product Details