Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-44
Entrez Gene ID
174210
Gene Name
protein-coding
Gene Aliases
CELE_C07D10.2
Chromosome Location
Chr.II: 7394955 - 7397351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063148.6 NP_495549.1 7 - 8 1042 145
NM_063149.5 NP_495550.1 7 - 8 1021 145

View Full Product Details