Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-7
Entrez Gene ID
182368
Gene Name
Zinc metalloproteinase nas-7
Gene Aliases
CELE_C07D10.4
Chromosome Location
Chr.II: 7400545 - 7402725 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063151.4 NP_495552.2 6 - 7 762 143

View Full Product Details