Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lir-2
Entrez Gene ID
174239
Gene Name
protein-coding
Gene Aliases
CELE_F18A1.4
Chromosome Location
Chr.II: 7667236 - 7671906 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026926.4 NP_001022097.1 2 - 3 104 104
GenBank mRNA AJ130959.1 - 1 - 2 59 104

View Full Product Details