The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026926.4 | NP_001022097.1 | 6 - 7 | 1015 | 124 |
NM_001026927.3 | NP_001022098.1 | 4 - 5 | 936 | 124 | |
GenBank mRNA | AJ130959.1 | - | 5 - 6 | 970 | 124 |
AJ130960.1 | - | 4 - 5 | 922 | 124 |