Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-4
Entrez Gene ID
174258
Gene Name
protein-coding
Gene Aliases
CELE_C06A8.9
Chromosome Location
Chr.II: 7785100 - 7794322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267197.2 NP_001254126.1 5 - 6 781 155
NM_001373747.3 NP_001359784.1 5 - 6 784 155

View Full Product Details