Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-7
Entrez Gene ID
174262
Gene Name
Acetylcholine receptor subunit alpha-type acr-7
Gene Aliases
CELE_T09A5.3
Chromosome Location
Chr.II: 7845631 - 7849797 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063246.5 NP_495647.1 9 - 10 1228 88
GenBank mRNA AF187012.1 - 9 - 10 1228 88

View Full Product Details