Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrf-6
Entrez Gene ID
174287
Gene Name
Nose resistant to fluoxetine protein 6
Gene Aliases
CELE_C08B11.4
Chromosome Location
Chr.II: 8028952 - 8032708 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063279.5 NP_495680.1 3 - 4 584 93
GenBank mRNA AF173372.1 - 3 - 4 584 93

View Full Product Details