Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ufd-2
Entrez Gene ID
174295
Gene Name
protein-coding
Gene Aliases
CELE_T05H10.5
Chromosome Location
Chr.II: 8062741 - 8067977 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383900.1 3 - 4 1304 78
NM_063290.3 NP_495691.1 3 - 4 1318 78

View Full Product Details