Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-2
Entrez Gene ID
174302
Gene Name
Serpentine receptor class epsilon-2
Gene Aliases
CELE_C41C4.2
Chromosome Location
Chr.II: 8100350 - 8102276 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063297.7 NP_495698.4 6 - 7 860 89

View Full Product Details