Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tub-1
Entrez Gene ID
174312
Gene Name
Tubby protein homolog 1
Gene Aliases
CELE_F10B5.4
Chromosome Location
Chr.II: 8154096 - 8156945 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063309.5 NP_495710.1 2 - 3 317 118
GenBank mRNA AF143297.1 - 2 - 3 337 118

View Full Product Details