Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rrf-3
Entrez Gene ID
174315
Gene Name
RNA-dependent RNA polymerase Family
Gene Aliases
CELE_F10B5.7
Chromosome Location
Chr.II: 8162465 - 8168651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063312.7 NP_495713.2 14 - 15 5195 82

View Full Product Details