Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-14
Entrez Gene ID
191601
Gene Name
AcetylCholine Receptor
Gene Aliases
CELE_T05C12.2
Chromosome Location
Chr.II: 8174937 - 8177195 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063315.3 NP_495716.1 4 - 5 355 107

View Full Product Details