Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-5
Entrez Gene ID
174317
Gene Name
protein-coding
Gene Aliases
CELE_T05C12.6
Chromosome Location
Chr.II: 8182323 - 8185109 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027145.3 NP_001022316.1 7 - 8 1446 96
NM_001027146.3 NP_001022317.1 7 - 8 1358 96
NM_001027147.5 NP_001022318.1 6 - 7 1237 96
GenBank mRNA AF063244.1 - 7 - 8 1380 96

View Full Product Details