Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-3
Entrez Gene ID
174321
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_M110.2
Chromosome Location
Chr.II: 8210584 - 8212780 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063326.3 NP_495727.1 3 - 4 618 73

View Full Product Details