Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tdc-1
Entrez Gene ID
174327
Gene Name
protein-coding
Gene Aliases
CELE_K01C8.3
Chromosome Location
Chr.II: 8270597 - 8273615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063342.6 NP_495743.1 7 - 8 1650 67
NM_063343.4 NP_495744.1 7 - 8 1660 67

View Full Product Details