Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nst-1
- Entrez Gene ID
-
174329
- Gene Name
- Guanine nucleotide-binding protein-like 3 homolog
- Gene Aliases
-
CELE_K01C8.9
- Chromosome Location
-
Chr.II: 8281142 - 8283208 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063348.6 |
NP_495749.1 |
2 - 3 |
139 |
93 |
- Gene Symbol
- mrpl-10
- Entrez Gene ID
-
186840
- Gene Name
- Mitochondrial Ribosomal Protein, Large
- Gene Aliases
-
CELE_K01C8.6
- Chromosome Location
-
Chr.II: 8277527 - 8283197 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063346.6 |
NP_495747.1 |
2 - 3 |
128 |
93 |
View Full Product Details