Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amt-3
Entrez Gene ID
174338
Gene Name
Putative ammonium transporter 3
Gene Aliases
CELE_M195.3
Chromosome Location
Chr.II: 8363172 - 8369159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063360.7 NP_495761.2 7 - 8 1465 82

View Full Product Details