Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubc-24
Entrez Gene ID
186057
Gene Name
UBIQUITIN_CONJUGAT_2 domain-containing protein
Gene Aliases
CELE_F49E12.4
Chromosome Location
Chr.II: 8405603 - 8406927 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063368.5 NP_495769.2 5 - 6 843 66

View Full Product Details