Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptf-2
Entrez Gene ID
188733
Gene Name
SP (Specificity Protein) Transcription Factor
Gene Aliases
CELE_T22C8.5
Chromosome Location
Chr.II: 8625459 - 8627622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063432.2 NP_495833.1 2 - 3 565 104

View Full Product Details